missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroglycan C/CSPG5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00€
Specifications
| Antigen | Neuroglycan C/CSPG5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Neuroglycan C/CSPG5 Polyclonal specifically detects Neuroglycan C/CSPG5 in Human samples. It is validated for Western Blot.Specifications
| Neuroglycan C/CSPG5 | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Neuroscience | |
| CALEB, chondroitin sulfate proteoglycan 5, chondroitin sulfate proteoglycan 5 (neuroglycan C), MGC44034, Neuroglycan C, NGCAcidic leucine-rich EGF-like domain-containing brain protein | |
| CSPG5 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| O95196 | |
| 10675 | |
| Synthetic peptides corresponding to CSPG5(chondroitin sulfate proteoglycan 5 (neuroglycan C)) The peptide sequence was selected from the N terminal of CSPG5. Peptide sequence LGGVTAEAGSGDAQALPATLQAPHEVLGQSIMPPAIPEATEASGPPSPTP. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title