missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98917-50ul
This item is not returnable.
View return policy
Description
Neurogenin-2 Polyclonal antibody specifically detects Neurogenin-2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Neurogenin-2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:200-1:1000, Immunoprecipitation 4-8 μL/mg of lysate | |
| Atoh4, BHLHA8, bHLHa8MGC46562, Class A basic helix-loop-helix protein 8, Math4A, neurogenin 2, neurogenin-2, ngn-2, NGN2, Protein atonal homolog 4 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Neurogenin-2. | |
| 50 μg | |
| Neuroscience, Signal Transduction | |
| 63973 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction