missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Netrin G2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94090-0.1ml
This item is not returnable.
View return policy
Description
Netrin G2 Polyclonal antibody specifically detects Netrin G2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Netrin G2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| bA479K20.1, KIAA0625, KIAA1857bA479K20.1 (novel protein), laminet 2, laminet-2, LHLL9381, LMNT2, MGC21884, netrin G1, netrin G2, netrin-G2, NTNG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 18-300 of human NTNG2 (NP_115925.2). DYDICKSWVTTDEGPTWEFYACQPKVMRLKDYVKVKVEPSGITCGDPPERFCSHENPYLCSNECDASNPDLAHPPRLMFDKEEEGLATYWQSITWSRYPSPLEANITLSWNKTVELTDDVVMTFEYGRPTVMVLEKSLDNGRTWQPYQFYAEDCMEAFGMSARRARDMSSSSAHRVLCTEEYSRWAGSKKEKHVRFEVRDRFAIFAGPDLRNMDNLYTRLESAKGLKEFFTLTDLRMRLLRPALGGTYVQRENLYKYFYAISNIEVIGRCKCNLHANLCSMRE | |
| 0.1 mL | |
| Neuroscience | |
| 84628 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction