missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NETO1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 624.00€
Specifications
| Antigen | NETO1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18201382
|
Novus Biologicals
NBP2-56508 |
100 μL |
624.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658118
|
Novus Biologicals
NBP2-56508-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NETO1 Polyclonal specifically detects NETO1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| NETO1 | |
| Polyclonal | |
| Rabbit | |
| Vision | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 81832 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PIIGRFCGQQNPPVIKSSGRFLWIKFFADGELESMGFSARYNFTPDPDFKD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 1, BTCL1BCTL1neuropilin and tolloid-like protein 1, FLJ41325, neuropilin (NRP) and tolloid (TLL)-like 1 | |
| NETO1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title