Description
- Tray that allows titre plates to be held securely and stacked vertically on the 1690 adjustable clamp as used with the MiniG homogeniser from SPEX SamplePrep.
- Tray to hold up two stacked deep well plates in 1690 clamp
Specifications
Specifications
| For Use With (Application) | Accessory for the SPEX MiniG homogeniser |