missing translation for 'onlineSavingsMsg'
Learn More

Nessler Reagent, Chem-Lab

Product Code. p-8014367
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11929232

Brand: Chem-Lab CL0214760100

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Used to detect ammonia. Nessler Reagent, Chem-Lab is a solution of potassium iodide and mercury chloride/mercury iodide, plus sodium hydroxide in water. It is used for qualitative determination of ammonia and ammonium compounds.

Nessler reagent (K2HgI4) is a solution prepared by combining sodium hydroxide and potassium iodide with mercuric chloride or mercury iodide. When reacted with gaseous ammonia, it produces brown fumes. When passed through a solution of ammonia, it produces a dirty brown precipitate.

Chemical Identifiers

CAS 7783-33-7
Molecular Formula H4HgI4K2
Molecular Weight (g/mol) 790.44
MDL Number MFCD00049656
InChI Key ZCPVFAXYZYZMBC-UHFFFAOYSA-N
Synonym nessler's reagent, nessler reagent, mayer's reagent, channing's solution, caswell no. 695, potassium iodomercurate, mercury ii potassium iodide, mercury potassium iodide, potassium mercuric iodide, mercuric potassium iodide
PubChem CID 24542
ChEBI CHEBI:51568
SMILES [K+].[K+].I.I.I.I.[Hg--]

Specifications

Color Yellow
Boiling Point 100°C
Melting Point 0°C
Composition 70g KI + 100g HgCl2 + 160g NaOH / l H2O
Solubility Information Water Soluble
IUPAC Name dipotassium tetraiodomercurydiuide
Formula Weight 786.40g/mol
Odor Odorless
pH >13
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Mercuric potassium iodide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nessler reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : KI,HgCl2,NaOH
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.