missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neprilysin-2/MMEL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-92125
This item is not returnable.
View return policy
Description
Neprilysin-2/MMEL1 Polyclonal antibody specifically detects Neprilysin-2/MMEL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Neprilysin-2/MMEL1 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| EC 3.4.24, EC 3.4.24.11, MELL1, membrane metallo-endopeptidase-like 1, Membrane metallo-endopeptidase-like 2MGC119455, MGC119454, MMEL2, NEP2, NEP2(m), NEPIIMGC119456, Neprilysin II, Neprilysin-2, NL2NL1, SEP, soluble secreted endopeptidase, zinc metallopeptidase | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LEELQSQFGLKGFNWTLFIQTVLSSVKIKLLPDEEVVVYGIPYLQNLENIIDTYSARTIQNYLVWRLVLDRIGSLSQRFKDTRVNYRKALFGTMVEEVRWRECVGYVNSNMENAVGSLYVREAFPGDSKSMVRELIDKVRTVFVETLD | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79258 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction