missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nefazodone Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Nefazodone Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Nefazodone Hydrochloride |
| CAS | 82752-99-6 |
| Analyte or Component Names | Nefazodone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H32 Cl N5 O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3H-1,2,4-Triazol-3-one, 2-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-5-ethyl-2,4-dihydro-4-(2-phenoxyethyl)-, hydrochloride (1:1)3H-1,2,4-Triazol-3-one, 2-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-5-ethyl-2,4-dihydro-4-(2-phenoxyethyl)-, monohydrochloride (9CI), BMY 13754, BMY 13754-1, Dutonin, MJ 13754-1, Nefazodone hydrochloride, Serazone, Serzone |
| InChI Formula | InChI=1S/C25H32ClN5O2.ClH/c1-2-24-27-31(25(32)30(24)18-19-33-23-10-4-3-5-11-23)13-7-12-28-14-16-29(17-15-28)22-9-6-8-21(26)20-22;/h3-6,8-11,20H,2,7,12-19H2,1H3;1H |
| SMILES | Cl.CCC1=NN(CCCN2CCN(CC2)c3cccc(Cl)c3)C(=O)N1CCOc4ccccc4 |
| IUPAC Name | 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-5-ethyl-4-(2-phenoxyethyl)-1,2,4-triazol-3-one;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction