missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDD4 Monoclonal Antibody (OTI5F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812643S
This item is not returnable.
View return policy
Description
NEDD4 Monoclonal antibody specifically detects NEDD4 in Human samples. It is validated for Western BlotSpecifications
| NEDD4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P46934 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4734 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5F8 | |
| Unconjugated | |
| NEDD4 | |
| AA959633; AL023035; AU019897; cell proliferation-inducing gene 53 protein; E3 ubiquitin-protein ligase NEDD4; E430025J12Rik; HECT-type E3 ubiquitin transferase NEDD4; Kiaa0093; mKIAA0093; NEDD4; NEDD-4; NEDD4 E3 ubiquitin protein ligase; Nedd4-1; Nedd4a; Neural precursor cell expressed developmentally down-regulated protein 4; neural precursor cell expressed, developmentally down-regulated 4; neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4; neural precursor cell expressed, developmentally down-regulated gene 4a; neural precursor cell expressed, developmentally down-regulted gene 4; OTTHUMP00000217840; OTTHUMP00000217841; OTTHUMP00000217842; PIG53; receptor-potentiating factor 1; RPF1 | |
| Human recombinant protein fragment corresponding to amino acids 160-460 of human NEDD4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction