missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-3/PVRL3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59519
This item is not returnable.
View return policy
Description
Nectin-3/PVRL3 Polyclonal specifically detects Nectin-3/PVRL3 in Human samples. It is validated for Western Blot.
Specifications
| Nectin-3/PVRL3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRR3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Guinea pig: 92%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NQS3 | |
| NECTIN3 | |
| Synthetic peptides corresponding to PVRL3(poliovirus receptor-related 3) The peptide sequence was selected from the N terminal of PVRL3. Peptide sequence SGKYICKAVTFPLGNAQSSTTVTVLVEPTVSLIKGPDSLIDGGNETVAAI. | |
| 100 μL | |
| Immunology | |
| 25945 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction