missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | NDUFV1 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232248
|
Novus Biologicals
NBP3-38496-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230263
|
Novus Biologicals
NBP3-38496-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NDUFV1 Polyclonal antibody specifically detects NDUFV1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| NDUFV1 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CI-51K, CI-51kD, CI51KD, complex I 51kDa subunit, complex I, mitochondrial respiratory chain, Complex I-51kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, FLJ59059, mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1, NADH dehydrogenase (ubiquinone) flavoprotein 1 (51kD), NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa, NADH dehydrogenase flavoprotein 1, NADH-ubiquinone oxidoreductase 51 kDa subunit, UQOR1NADH dehydrogenase [ubiquinone] flavoprotein 1, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 72-420 of human NDUFV1 (NP_009034.2).,, Sequence:, GPDWILGEIKTSGLRGRGGAGFPTGLKWSFMNKPSDGRPKYLVVNADEGEPGTCKDREILRHDPHKLLEGCLVGGRAMGARAAYIYIRGEFYNEASNLQVAIREAYEAGLIGKNACGSGYDFDVFVVRGAGAYICGEETALIESIEGKQGKPRLKPPFPADVGVFGCPTTVANVETVAVSPTICRRGGTWFAGFGRERNSGTKLFNISGHVNHPCTVEEEMSVPLKELIEKHAGGVTGGWDNLLAVIPGGSSTPLIPKSVCETVLMDFDALVQAQTGLGTAAVIVMDRSTDIVKAIARLIEFYKHESCGQCTPCREGVDWMNKVMARFVRGDARPAEIDSLWEISKQIE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 4723 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title