missing translation for 'onlineSavingsMsg'
Learn More

NDUFAF4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

415.00€

Specifications

Antigen NDUFAF4
Dilution Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18408661
View Documents
Novus Biologicals
NBP1-92171-25ul
25 μL
415.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NDUFAF4 Polyclonal specifically detects NDUFAF4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

NDUFAF4
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
29078
This antibody was developed against Recombinant Protein corresponding to amino acids:IKSIPKGKISIVEALTLLNNHKLFPETWTAEKIMQEYQLEQKDVNSLLKYFVTFEVEIFPPEDKKAIRSK
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Rabbit
Human
bA22L21.1, C6orf66chromosome 6 open reading frame 66, Hormone-regulated proliferation-associated protein of 20 kDa, hormone-regulated proliferation-associated protein, 20 kDa, HRPAP20HSPC125, My013, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 4
NDUFAF4
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.