missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFAF2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41116-0.025mg
This item is not returnable.
View return policy
Description
NDUFAF2 Polyclonal specifically detects NDUFAF2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence.
Specifications
| NDUFAF2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| NDUFAF2 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human Mimitin. The immunogen is located within the last 50 amino acids of Mimitin. Amino Acid Squence: PGSWMPRDGKSHNQ | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| B17.2LFLJ22398, B17.2-like, mimitin, MMTNmitochondrial, Myc-induced mitochondrial protein, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 2, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2, NDUFA12-like, NDUFA12-like protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 91942 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction