missing translation for 'onlineSavingsMsg'
Learn More

NDUFA2 Monoclonal Antibody (OTI5A5), TrueMAB™, OriGene

Product Code. 17585464
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17585464 30 μL 30µL
17565464 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17585464 Supplier Origene Technologies Supplier No. TA811465S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NDUFA2 Monoclonal antibody specifically detects NDUFA2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen NDUFA2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI5A5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NEMF
Gene Accession No. O60524
Gene Alias AV000592; B8; C1-B8; CCDC76; CD14; CIB8; CI-B8; complex I B8 subunit; complex I-B8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2; NADH:ubiquinone oxidoreductase subunit A2; NADH-ubiquinone oxidoreductase B8 subunit; NADH-ubiquinone oxidoreductase subunit CI-B8; NDUFA2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 869-1076 of human NEMF produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9147
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.