missing translation for 'onlineSavingsMsg'
Learn More

NCOA4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

369.00€ - 560.70€

Specifications

Antigen NCOA4
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18263405
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-56237
100 μL
ONLINE EXCLUSIVE PRICE
593.00€
560.70€ / 100µL
Save 32.30€
5% Off
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
18697118
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-56237-25ul
25 μL
369.00€
25µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NCOA4 Polyclonal specifically detects NCOA4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

NCOA4
Polyclonal
Rabbit
Human
Androgen receptor coactivator 70 kDa protein, Androgen receptor-associated protein of 70 kDa, ARA70RFGret fused, DKFZp762E1112, ELE1NCoA-4, nuclear receptor coactivator 4,70 kDa AR-activator, PTC3RET-activating gene ELE1, Ret-activating protein ELE1,70 kDa androgen receptor coactivator
NCOA4
IgG
Affinity Purified
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
8031
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KSPMNTSWCSFNTADWVLPGKKMGNLSQLSSGEDKWLLRKKAQEVLLNSPLQEEHNFPPDHYGLPAVCDLFACMQLKVDKEKWLYRTPLQ
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.