missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nateglinide EP Impurity B, Mikromol™
Description
Nateglinide EP Impurity B
Specifications
Specifications
| Chemical Name or Material | N-[(trans-4-(1-Methylethyl)cyclohexyl)carbonyl]-L-phenylalanine |
| CAS | 105816-05-5 |
| Analyte or Component Names | N-[(trans-4-(1-Methylethyl)cyclohexyl)carbonyl]-L-phenylalanine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H27 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-L-phenylalanine (Nateglinide L-Phenylalanine Isomer), Nateglinide Imp. B (EP), Nateglinide USP Related Compound B, Nateglinide USP RC B, N-[[trans-4-(Propan-2-yl)cyclohexyl]carbonyl]-L-phenylalanine, Nateglinide Imp. B (Pharmeuropa), N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-L-phenylalanine, Nateglinide L-Phenylalanine Isomer, Nateglinide Related Compound B, Nateglinide Impurity B |
| InChI Formula | InChI=1S/C19H27NO3/c1-13(2)15-8-10-16(11-9-15)18(21)20-17(19(22)23)12-14-6-4-3-5-7-14/h3-7,13,15-17H,8-12H2,1-2H3,(H,20,21)(H,22,23)/t15-,16-,17-/m0/s1 |
| SMILES | CC(C)[C@@H]1CC[C@H](CC1)C(=O)N[C@@H](Cc2ccccc2)C(=O)O |
| IUPAC Name | (2S)-3-phenyl-2-[(4-propan-2-ylcyclohexanecarbonyl)amino]propanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?