missing translation for 'onlineSavingsMsg'
Learn More

NARS Antibody, R&D Systems™

Product Code. 30229642 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30229642 100 μL 100µL
30228449 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30229642 Supplier Novus Biologicals Supplier No. NBP335181100ul
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

NARS Polyclonal specifically detects NARS in Human∣Mouse samples. It is validated for ELISA∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NARS
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias ASNRS, asparagine tRNA ligase 1, cytoplasmic, Asparagine--tRNA ligase, asparaginyl-tRNA synthetase, asparaginyl-tRNA synthetase, cytoplasmic, EC 6.1.1.22, NARS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 309-548 of human NARS (NP_004530.1).,, Sequence:, PALGDVFCIAQSYRAEQSRTRRHLAEYTHVEAECPFLTFDDLLNRLEDLVCDVVDRILKSPAGSIVHELNPNFQPPKRPFKRMNYSDAIVWLKEHDVKKEDGTFYEFGEDIPEAPERLMTDTINEPILLCRFPVEIKSFYMQRCPEDSRLTESVDVLMPNVGEIVGGSMRIFDSEEILAGYKREGIDPTPYYWYTDQRKYGTCPHGGYGLGLERFLTWILNRYHIRDVCLYPRFVQRCTP
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 4677
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.