missing translation for 'onlineSavingsMsg'
Learn More

CD154 Mouse anti-Human, BUV737, Clone: TRAP1 (also known as TRAP-1), BD Biosciences™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 748983

Product Code. 17220565

  • ONLINE EXCLUSIVE PRICE
    540.00€
    539.91€ / 50µg
    Save 0.09€
    0.02% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD154 Monoclonal antibody specifically detects CD154 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD154
Monoclonal
Brilliant Ultraviolet 737
CD40LG; CD40 ligand; CD40L; TNFSF5; T-BAM; TRAP
Human TRAP1 Transfected Cell Line
RUO
Human
IgG1 κ
Flow Cytometry
TRAP1 (also known as TRAP-1)
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.