missing translation for 'onlineSavingsMsg'
Learn More

Naproxen Antibody, Novus Biologicals™

Product Code. 18243816 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18243816 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18243816 Supplier Novus Biologicals Supplier No. NBP288855

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Naproxen Polyclonal specifically detects Naproxen in Non-species specific samples. It is validated for ELISA, Immunoaffinity Purification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Naproxen
Applications ELISA, Affinity Purification
Classification Polyclonal
Concentration 0.8 mg/ml
Conjugate Unconjugated
Dilution ELISA 1:300, Immunoaffinity Purification
Formulation Whole antiserum
Gene Alias (+)-Naproxen, (S)-2-(6-methoxynaphthalen-2-yl)propanoic acid, (S)-Naproxen, 22204-53-1, Equiproxen, Naprosyn, Naproxene
Host Species Rabbit
Immunogen BSA-Naproxen derivative conjugate
Purification Method Caprylic acid precipitation
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.