missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPG Antibody (4B5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00008774-M03
This item is not returnable.
View return policy
Description
NAPG Monoclonal antibody specifically detects NAPG in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| NAPG | |
| Monoclonal | |
| Unconjugated | |
| NP_003817 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 4B5 | |
| In 1x PBS, pH 7.4 | |
| gamma SNAP, GAMMASNAP, gamma-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein gamma, N-ethylmaleimide-sensitive factor attachment protein, gamma, SNAPG, SNAP-gamma, soluble NSF attachment protein | |
| NAPG (NP_003817.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAQKINEGLEHLAKAEKYLKTGFLKWKPDYDSAASEYGKAAVAFKNAKQFEQAKDACLREAVAHENNRALFHAAKAYEQAGMMLKEMQKLPEAVQLIEK | |
| 0.1 mg | |
| Neuroscience, Signal Transduction | |
| 8774 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction