missing translation for 'onlineSavingsMsg'
Learn More

Nanos3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

178.00€

Specifications

Antigen Nanos3
Concentration 1 mg/mL
Dilution Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18593243
View Documents
Novus Biologicals
NBP1-76919-0.025mg
0.025 mg
178.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nanos3 Polyclonal specifically detects Nanos3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Nanos3
Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
Polyclonal
Rabbit
Human
P60323
342977
Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of the human Nanos3. The immunogen is located within the last 50 amino acids of Nanos3. Amino Acid Squence: CKLVRPDKAKQDTGHR
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
20 kDa
1 mg/mL
Western Blot
Unconjugated
RUO
PBS with 0.02% Sodium Azide
MGC120114, nanos homolog 3, nanos homolog 3 (Drosophila), NOS-3, NOS3NANOS1L
NANOS3
IgG
Affinity Purified
This Nanos3 antibody will not cross-react with either Nanos 1 or Nanos2.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.