missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NANOS2 Monoclonal Antibody (343255)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561217
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
NANOS2 is a gene that encodes for a protein found in the testes and ovaries. It is a member of the NANOS family of proteins and plays a crucial role in the regulation of germ cell development. NANOS2 is involved in the maintenance of germ cell pluripotency and the suppression of somatic cell differentiation. It is also involved in the regulation of meiosis and the formation of gametes. Mutations in the NANOS2 gene have been associated with various reproductive disorders, including infertility and testicular germ cell tumors. NANOS2 is a target of research in the fields of reproductive biology and genetics.
Specifications
| NANOS2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P60321 | |
| NANOS2 | |
| E. coli-derived recombinant human Nanos2, Met1-Arg138. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 343255 | |
| Unconjugated | |
| NANOS2 | |
| nanos C2HC-type zinc finger 2; nanos homolog 2; nanos homolog 2 (Drosophila); NANOS2; nos2; NOS-2; RGD1562436; ZC2HC12B | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 339345 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction