missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanog Antibody (CL5803) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61428H
This item is not returnable.
View return policy
Description
Nanog Monoclonal antibody specifically detects Nanog in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Nanog | |
| Monoclonal | |
| HRP | |
| FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
| This antibody was developed using a synthetic peptide derived from Q9H9S0, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Cellular Markers, Macroautophagy, mTOR Pathway, Neurodegeneration, Protein Turnover, Regulatory Immunology, Signal Transduction, Stem Cell Markers, Stem Cell Signaling Pathway, Ubiquitin Proteasome Pathway | |
| 79923 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL5803 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction