Learn More
Macherey-Nagel™ Nano-SIL NH2/UV254 HPTLC Plate
Product Code. 10259791
Change view
Click to view available options
Dimensions (L x W):
10 x 10 cm
10 x 20 cm
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10259791 10 x 20 cm Pack of 25
10187401 10 x 10 cm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10259791 Supplier Macherey-Nagel™ Supplier No. 811112
Only null left
Specifications
Material Nano-SIL NH2
Particle Size 2 to 10 μm
Pore Size 60 Å
Quantity 25 Pk.
Type HPTLC Plate
Fluorescent Indicator UV254
For Use With (Application) Vitamins, Sugars, Steroids, Purine Derivatives, Xanthines, Phenols, Nucleotides and Pesticides
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.2 mm
Dimensions (L x W) 10 x 20 cm