Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 555, Chromotek
Product Code. 18034245
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18034245 10 μL 10µL
18094725 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18034245 Supplier Chromotek Supplier No. shuGCL555210
 Restricted Item
In Stock
Add to basket
Add to basket
Alpaca Recombinant Monoclonal Antibody
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Specifications
Antigen Nano-Secondary™
Conjugate CoraLite Plus 555
Host Species Alpaca
Quantity 10 μL
Primary or Secondary Secondary
Target Species Human
Form Liquid
Isotype IgG