missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naltrexone Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Naltrexone hydrochloride |
| CAS | 16676-29-2 |
| Analyte or Component Names | Naltrexone hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H23 N O4 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Antaxone, Depade, EN 1639A, NIH 8503, Nalorex, Naltrexone hydrochloride, ReVia, Trexan, (5alpha)-17-(Cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-one Hydrochloride (1:1), Morphinan-6-one, 17-(cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxy-, hydrochloride, (5alpha)- (9CI), Morphinan-6-one, 17-(cyclopropylmethyl)-4,5alpha-epoxy-3,14-dihydroxy-, hydrochloride (8CI) |
| InChI Formula | InChI=1S/C20H23NO4.ClH/c22-13-4-3-12-9-15-20(24)6-5-14(23)18-19(20,16(12)17(13)25-18)7-8-21(15)10-11-1-2-11;/h3-4,11,15,18,22,24H,1-2,5-10H2;1H/t15-,18+,19+,20-;/m1./s1 |
| Solution Type | Neat |
| SMILES | Cl.Oc1ccc2C[C@H]3N(CC4CC4)CC[C@@]56[C@@H](Oc1c25)C(=O)CC[C@@]36O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction