missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NALP12 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
610.00€
Specifications
| Antigen | NALP12 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Form | Purified |
Description
NALP12 Polyclonal antibody specifically detects NALP12 in Human, Mouse, Rat, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| NALP12 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FCAS2, Monarch-1, NACHT, leucine rich repeat and PYD containing 12, NACHT, LRR and PYD domains-containing protein 12, NALP12LRR and PYD containing protein 12, NLR family, pyrin domain containing 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 12, PYPAF7Monarch1, PYRIN-containing APAF1-like protein 7, Regulated by nitric oxide, RNO2 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human NALP12. The immunogen is located within amino acids 110 - 160 of NALP12. Amino Acid Squence: TPRKDPQETYRDYVRRKF | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| PerCP | |
| Rabbit | |
| Apoptosis | |
| PBS | |
| 91662 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title