missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naftazone, LoGiCal
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Naftazone
Specifications
Specifications
| Chemical Name or Material | Naftazone |
| CAS | 15687-37-3 |
| Analyte or Component Names | Naftazone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H9 N3 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-(1-Oxo-2(1 H)-naphthalenylidene)-hydrazinecarboxamide, Hydrazinecarboxamide, 2-(1-oxo-2(1 H)-naphthalenylidene)-, 1,2-Naphthoquinone, 2-semicarbazone (8 CI), 1,2-Naphthaquinone 2-semicarbazone, Etioven, Haemostop, Haemostop Injection, Karbinon, Karbinone, Mediaven, Naftazon, Naftazone |
| InChI Formula | InChI=1 S/C11H9N3O2/c12-11(16)14-13-9-6-5-7-3-1-2-4-8(7)10(9)15/h1-6 H,(H3,12,14,16)/b13-9+ |
| SMILES | NC(=O)N\N=C\1/C=Cc2ccccc2C1=O |
| IUPAC Name | [(E)-(1-oxonaphthalen-2-ylidene)amino]urea |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction