Learn More
Nafronyl, TRC
Product Code. 30005951
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005951 5 mg 5mg
30005926 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30005951 Supplier Toronto Research Chemicals Supplier No. TRCN2150005MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31329-57-4
Molecular Formula C24H33NO3
Molecular Weight (g/mol) 383.52
Synonym 2-Furanpropanoic acid, tetrahydro-α-(1-naphthalenylmethyl)-, 2-(diethylamino)ethyl ester, 2-Furanpropionic acid, tetrahydro-α-(1-naphthylmethyl)-, 2-(diethylamino)ethyl ester (7CI,8CI), 2-(Diethylamino)ethyl tetrahydro-α-(1-naphthylmethyl)-2-furanpropionate ester, Gevatran, LS 84, N-Diethylamino ethyl β-(1-naphthol)-β-(2-tetrahydrofuryl)isobutyrate, Nafronyl, Naftidrofuryl, Naphtidrofuryl, Tridus
SMILES CCN(CC)CCOC(=O)C(CC1CCCO1)Cc2cccc3ccccc23
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C24H33NO3/c1-3-25(4-2)14-16-28-24(26)21(18-22-12-8-15-27-22)17-20-11-7-10-19-9-5-6-13-23(19)20/h5-7,9-11,13,21-22H,3-4,8,12,14-18H2,1-2H3
Formula Weight 383.246
Percent Purity >95%
Chemical Name or Material Nafronyl
Grade HPLC