CAS RN 31329-57-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nafronyl, TRC
CAS: 31329-57-4 Molecular Formula: C24H33NO3 Molecular Weight (g/mol): 383.52 Synonym: 2-Furanpropanoic acid, tetrahydro-α-(1-naphthalenylmethyl)-, 2-(diethylamino)ethyl ester,2-Furanpropionic acid, tetrahydro-α-(1-naphthylmethyl)-, 2-(diethylamino)ethyl ester (7CI,8CI),2-(Diethylamino)ethyl tetrahydro-α-(1-naphthylmethyl)-2-furanpropionate ester,Gevatran,LS 84,N-Diethylamino ethyl β-(1-naphthol)-β-(2-tetrahydrofuryl)isobutyrate,Nafronyl,Naftidrofuryl,Naphtidrofuryl,Tridus SMILES: CCN(CC)CCOC(=O)C(CC1CCCO1)Cc2cccc3ccccc23