missing translation for 'onlineSavingsMsg'
Learn More

N6-methyladenosine (m6A) Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18627427 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18627427 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18627427 Supplier Novus Biologicals Supplier No. NBP305657

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

N6-methyladenosine (m6A) Polyclonal antibody specifically detects N6-methyladenosine (m6A) in Broad Species Reactivity samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Dot Blot, Microarray, RNA immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen N6-methyladenosine (m6A)
Applications Immunofluorescence, Immunoprecipitation, Dot Blot, Microarray, RNA Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200, Dot Blot 1:500 - 1:2000, Microarray 1:50 - 1:200, RNA immunoprecipitation 1:50 - 1:200
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine
Host Species Rabbit
Immunogen Chemical compounds corresponding to N6-methyladenosine (m6A).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics, metabolism
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.