missing translation for 'onlineSavingsMsg'
Learn More

N4BP2L2 Monoclonal Antibody (OTI3F12), TrueMAB™, OriGene

Product Code. 17573653
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17573653 30 μL 30µL
17553653 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17573653 Supplier Origene Technologies Supplier No. TA804204S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

N4BP2L2 Monoclonal antibody specifically detects N4BP2L2 in Human samples. It is validated for Western Blot

Specifications

Antigen N4BP2L2
Applications Western Blot
Classification Monoclonal
Clone OTI3F12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene N4BP2L2
Gene Accession No. Q92802
Gene Alias 2700092H06Rik; 92M18.3; CG005; CG016; N4BP2L2; NEDD4 binding protein 2 like 2; NEDD4 binding protein 2-like 2; NEDD4-binding protein 2-like 2; PFAAP5; phosphonoformate immuno-associated protein 5; Phosphonoformate immuno-associated protein 5 homolog; protein from BRCA2 region; zag1
Host Species Mouse
Immunogen Full length human recombinant protein of human N4BP2L2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10443
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.