Learn More
N-[Tris(hydroxymethyl)methyl]acrylamide, 99%
Product Code. 11469887
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469887 5 g 5g
11479887 25 g 25g
11489887 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11469887 Supplier Thermo Scientific Alfa Aesar Supplier No. B22854.06
Request Bulk or Custom Format
CAS: 13880-05-2 | C7H13NO4 | 175.18 g/mol
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.
Solubility
Soluble in water.
Notes
Hygroscopic. Light sensitive. Incompatible with strong oxdizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13880-05-2
Molecular Formula C7H13NO4
Molecular Weight (g/mol) 175.18
MDL Number MFCD00081045
InChI Key MVBJSQCJPSRKSW-UHFFFAOYSA-N
Synonym n-tris hydroxymethyl methyl acrylamide, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl acrylamide, tris-acrylamide, n-acryloyltris hydroxymethyl aminomethane, n-acryloyl-tris hydroxymethyl aminomethane, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl prop-2-enamide, zlchem 121, acryloylaminomethylidynetrimethanol, n-1,3-dihydroxy-2-hydroxymethyl-2-propanyl acrylamide
PubChem CID 83788
IUPAC Name N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide
SMILES OCC(CO)(CO)NC(=O)C=C
Specifications
Melting Point 134°C to 138°C
Linear Formula CH2=CHCONHC(CH2OH)3
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 175.18
Percent Purity 99%
Chemical Name or Material N-[Tris(hydroxymethyl)methyl]acrylamide
compliance-icons
Product Identifier
  • N-Acryloyltris(hydroxymethyl)aminomethane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: N-Acryloyltris(hydroxymethyl)aminomethane