Learn More
N-Succinyl-5-aminoimidazole-4-carboxamide Ribose 5’-Phosphate, TRC
Product Code. 30012219
Change view
Click to view available options
Quantity:
5 mg
0.5 mL
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012219 5 mg 5mg
30011311 0.5 mL 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30012219 Supplier Toronto Research Chemicals Supplier No. TRCS6887955MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3031-95-6
Molecular Formula C13H19N4O12P
Molecular Weight (g/mol) 454.28
Synonym N-[[5-Amino-1-(5-O-phosphono-β-D-ribofuranosyl)-1H-imidazol-4-yl]carbonyl]-L-aspartic Acid, N-[(5-Amino-1-β-D-ribofuranosylimidazol-4-yl)carbonyl]-
IUPAC Name (5-amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-((phosphonooxy)methyl)tetrahydrofuran-2-yl)-1H-imidazole-4-carbonyl)-L-aspartic acid
SMILES NC(N([C@H]1[C@H](O)[C@H](O)[C@@H](COP(O)(O)=O)O1)C=N2)=C2C(N[C@H](C(O)=O)CC(O)=O)=O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C13H19N4O12P/c14-10-7(11(22)16-4(13(23)24)1-6(18)19)15-3-17(10)12-9(21)8(20)5(29-12)2-28-30(25,26)27/h3-5,8-9,12,20-21H,1-2,14H2,(H,16,22)(H,18,19)(H,23,24)(H2,25,26,27)/t4-,5+,8+,9+,12+/m0/s1
Formula Weight 454.07
Percent Purity >95%
Chemical Name or Material N-Succinyl-5-aminoimidazole-4-carboxamide Ribose 5'-Phosphate
Grade HPLC