CAS RN 3031-95-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Succinyl-5-aminoimidazole-4-carboxamide Ribose 5’-Phosphate, TRC
CAS: 3031-95-6 Molecular Formula: C13H19N4O12P Molecular Weight (g/mol): 454.28 Synonym: N-[[5-Amino-1-(5-O-phosphono-β-D-ribofuranosyl)-1H-imidazol-4-yl]carbonyl]-L-aspartic Acid,N-[(5-Amino-1-β-D-ribofuranosylimidazol-4-yl)carbonyl]- IUPAC Name: (5-amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-((phosphonooxy)methyl)tetrahydrofuran-2-yl)-1H-imidazole-4-carbonyl)-L-aspartic acid SMILES: NC(N([C@H]1[C@H](O)[C@H](O)[C@@H](COP(O)(O)=O)O1)C=N2)=C2C(N[C@H](C(O)=O)CC(O)=O)=O