missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam N-Palmitoyl Phosphatidylethanolamine, Endogenous Lipid Anorectic Hormone, 1 mg
Description
- Molecular Weight: 930.4 g/mol
- Purity: >98%
- CAS Number: 108321-22-8
- Storage Temperature: Store at -20°C
- Shipping Temperature: Ambient - Can Ship with Ice
- Composition: 100% N-Palmitoyl phosphatidylethanolamine
- Biological Activity: Endogenous lipid anorectic hormone that reduces food intake and body weight in rats and mice, acting via the CNS
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% N-Palmitoyl phosphatidylethanolamine |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Lipid-based Substrate |
| Target Molecule | N-Palmitoyl Phosphatidylethanolamine |
| Formulation | 100% N-Palmitoyl phosphatidylethanolamine |
| Quantity | 1 mg |
| For Use With (Application) | Obesity-treatment research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?