missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-PAC Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93036-0.02ml
This item is not returnable.
View return policy
Description
N-PAC Polyclonal antibody specifically detects N-PAC in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| N-PAC | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BM045, Cytokine-like nuclear factor N-PAC, Glyoxylate reductase 1 homolog, glyoxylate reductase 1 homolog (Arabidopsis), HIBDLnuclear protein 60 kDa, NP603-hydroxyisobutyrate dehydrogenase-like protein, N-PAC, nuclear protein 60kDa, Nuclear protein NP60, Nuclear protein of 60 kDa, putative oxidoreductase GLYR1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-270 of human GLYR1 (NP_115958.2). MIKINKGKRFQQAVDAVEEFLRRAKGKDQTSSHNSSDDKNRRNSSEERSRPNSGDEKRKLSLSEGKVKKNMGEGKKRVSSGSSERGSKSPLKRAQEQSPRKRGRPPKDEKDLTIPESSTVKGMMAGPMAAFKWQPTASEPVKDADPHFHHFLLSQTEKPAVCYQAITKKLKICEEETGSTSIQAADSTAVNGSITPTDKKI | |
| 0.02 mL | |
| Signal Transduction | |
| 84656 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction