missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosonornicotine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | (±)-N'-Nitrosonornicotine (NNN) |
| CAS | 80508-23-2 |
| Analyte or Component Names | (±)-N'-Nitrosonornicotine (NNN) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H11 N3 O |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | N-Nitrosonornicotine, rac-N'-Nitroso Nornicotine |
| InChI Formula | InChI=1S/C9H11N3O/c13-11-12-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5,7,9H,2,4,6H2 |
| SMILES | O=NN1CCCC1c2cccnc2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction