Learn More
N-Nitrosodimethylamine (10 mg/mL in Ethanol), TRC
Product Code. 30003738
Change view
Click to view available options
Quantity:
1 x 5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003738 1 x 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30003738 Supplier Toronto Research Chemicals Supplier No. TRCKIT91001X5ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 62-75-9
Analyte or Component Names N-Nitrosodimethylamine
Shipping Condition Room Temperature
Molecular Formula C2 H6 N2 O
Concentration 10 mg/mL
Synonym Methanamine, N-methyl-N-nitroso-, Dimethylamine, N-nitroso- (8CI), N-Methyl-N-nitrosomethanamine, DMN, DMNA, Dimethylnitrosamine, N-Nitroso-N,N-dimethylamine, N-Nitrosodimethylamine, NDMA, NSC 23226, N-Nitroso-dimethylamine
InChI Formula InChI=1S/C2H6N2O/c1-4(2)3-5/h1-2H3
SMILES CN(C)N=O
IUPAC Name N,N-dimethylnitrous amide
Solvent or Matrix Ethanol
Molecular Weight (g/mol) 74.08
Quantity 1 x 5 mL
Formula Weight 74.048
Concentration or Composition (by Analyte or Components) N-Nitrosodimethylamine 10 mg/mL
Chemical Name or Material N-Nitrosodimethylamine