Learn More
N-Nitrosoanabasine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30320882
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320882 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30320882 Supplier Dr. Ehrenstorfer Supplier No. DREA15600500AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (RS)-N-Nitrosoanabasine
CAS 37620-20-5
Analyte or Component Names (RS)-N-Nitrosoanabasine
Shipping Condition Room Temperature
Molecular Formula C10 H13 N3 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Pyridine, 3-(1-nitroso-2-piperidinyl)-, Piperidine, 1-nitroso-2-(3-pyridyl)- (6CI,7CI), 3-(1-Nitroso-2-piperidinyl)pyridine, (R,S)-N-Nitroso Anabasine, (RS)-N-Nitrosoanabasine
InChI Formula InChI=1S/C10H13N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h3-4,6,8,10H,1-2,5,7H2
SMILES O=NN1CCCCC1c2cccnc2
IUPAC Name 3-(1-nitrosopiperidin-2-yl)pyridine
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 191.23
Quantity 1 mL
Formula Weight 191.1059 g/mol