Learn More
N-Nitroso Mirabegron, TRC
Product Code. 30427876
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427876 100 mg 100mg
30425194 10 mg 10mg
30420257 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30427876 Supplier Toronto Research Chemicals Supplier No. TRCN703565100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Nitroso Mirabegron
CAS 3017159-61-1
Analyte or Component Names N-Nitroso Mirabegron
Shipping Condition Room Temperature
Molecular Formula C21 H23 N5 O3 S
Synonym (R)-2-(2-aminothiazol-4-yl)-N-(4-(2-((2-hydroxy-2-phenylethyl)(nitroso)amino)ethyl)phenyl)acetamide
InChI Formula InChI=1S/C21H23N5O3S/c22-21-24-18(14-30-21)12-20(28)23-17-8-6-15(7-9-17)10-11-26(25-29)13-19(27)16-4-2-1-3-5-16/h1-9,14,19,27H,10-13H2,(H2,22,24)(H,23,28)/t19-/m0/s1
Solution Type Neat
SMILES O=C(NC1=CC=C(CCN(C[C@H](O)C2=CC=CC=C2)N=O)C=C1)CC3=CSC(N)=N3
IUPAC Name (R)-2-(2-aminothiazol-4-yl)-N-(4-(2-((2-hydroxy-2-phenylethyl)(nitroso)amino)ethyl)phenyl)acetamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 425.51
Quantity 100 mg
Formula Weight 425.15