Learn More
N,N'-(Oxydibenzene-4,1-diyl)diacetamide, Mikromol™
Product Code. 17981406
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17981406 100 mg 100mg
17962626 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17981406 Supplier Mikromol Supplier No. MM0042.20
 Restricted Item
Estimated Shipment: 13-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N,N'-(Oxydibenzene-4,1-diyl)diacetamide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N,N'-(Oxydibenzene-4,1-diyl)diacetamide
CAS 3070-86-8
Analyte or Component Names N,N'-(Oxydibenzene-4,1-diyl)diacetamide
Shipping Condition Room Temperature
Molecular Formula C16 H16 N2 O3
Certifications/Compliance ISO 17025
Synonym N,N'-(oxydi-4,1-phenylene)bisacetamide, Acetanilide, 4',4''-oxybis- (6CI,7CI,8CI), N,N'-(Oxydi-4,1-phenylene)bis[acetamide], 4,4'-Bis(acetylamino)diphenyl ether, 4,4'-Diacetamidodiphenyl ether, 4,4'-Oxybis[acetanilide], 4',4''-Oxybisacetanilide, Bis(p-acetylaminophenyl) ether, NSC 19584, N,N′-(Oxydibenzene-4,1-diyl)diacetamide, Paracetamol Imp. N (Pharmeuropa), Paracetamol Impurity N
InChI Formula InChI=1S/C16H16N2O3/c1-11(19)17-13-3-7-15(8-4-13)21-16-9-5-14(6-10-16)18-12(2)20/h3-10H,1-2H3,(H,17,19)(H,18,20)
SMILES CC(=O)Nc1ccc(Oc2ccc(NC(=O)C)cc2)cc1
IUPAC Name N-[4-(4-acetamidophenoxy)phenyl]acetamide
Molecular Weight (g/mol) 284.31
Quantity 100 mg
Formula Weight 284.1161 g/mol