Learn More
N,N'-Di-n-butylurea, 98%
Product Code. 11312406
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312406 25 g 25g
11302406 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11312406 Supplier Thermo Scientific Alfa Aesar Supplier No. L06165.14
Request Bulk or Custom Format
CAS: 1792-17-2 | C9H20N2O | 172.27 g/mol
N,N'-Di-n-butylurea is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Di-n-butylurea is used in agrochemical, pharmaceutical and dyestuff field.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1792-17-2
Molecular Formula C9H20N2O
Molecular Weight (g/mol) 172.27
MDL Number MFCD00015229
InChI Key AQSQFWLMFCKKMG-UHFFFAOYSA-N
Synonym n,n'-dibutylurea, n,n'-di-n-butylurea, urea, n,n'-dibutyl, 1,3-dibutyl urea, unii-j2hug1i10w, j2hug1i10w, n-butyl butylamino carboxamide, nn'-dibutylurea, urea,n'-dibutyl, 1,3-dibutyl-urea
PubChem CID 74523
IUPAC Name 1,3-dibutylurea
SMILES CCCCNC(=O)NCCCC
Specifications
Melting Point 69°C to 75°C
Boiling Point 115°C (0.03 mmHg)
Quantity 25 g
Beilstein 1704165
Formula Weight 172.27
Percent Purity 98%
Chemical Name or Material N,N'-Di-n-butylurea