accessibility menu, dialog, popup

UserName

CAS RN 1792-17-2

H3CHNOHNCH3

CAS RN 1792-17-2

IUPAC Name: 1,3-dibutylurea
Synonyms: 1,3-dibutylurea
Molecular Weight (g/mol): 172.27
Molecular Formula: C9H20N2O
InChi Key: AQSQFWLMFCKKMG-UHFFFAOYSA-N
SMILES: CCCCNC(=O)NCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results