missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamide, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
N,N'-Bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamide
Specifications
Specifications
| Chemical Name or Material | N,N'-bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamide |
| CAS | 76820-34-3 |
| Analyte or Component Names | N,N'-bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H19 N3 O8 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N,N'-bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamide, Iohexol Related Compound C |
| InChI Formula | InChI=1S/C14H19N3O8/c18-6-11(20)4-15-13(22)8-1-9(3-10(2-8)17(24)25)14(23)16-5-12(21)7-19/h1-3,11-12,18-21H,4-7H2,(H,15,22)(H,16,23) |
| SMILES | OCC(O)CNC(=O)c1cc(cc(c1)[N+](=O)[O-])C(=O)NCC(O)CO |
| IUPAC Name | 1-N,3-N-bis(2,3-dihydroxypropyl)-5-nitrobenzene-1,3-dicarboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction