missing translation for 'onlineSavingsMsg'
Learn More

n-Myc Mouse anti-Human, Mouse, Rat, Clone: NMYC-1, Novus Biologicals™

Mouse Monoclonal Antibody

484.00€

Specifications

Antigen n-Myc
Clone NMYC-1
Concentration 0.2 mg/mL
Dilution Western Blot 1 to 2 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL
Applications Western Blot, Immunofluorescence
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18694309
View Documents
Novus Biologicals
NBP3-07393-100ug
100 μg
484.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

n-Myc Monoclonal specifically detects n-Myc in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

n-Myc
0.2 mg/mL
Western Blot, Immunofluorescence
Unconjugated
Mouse
Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
10 mM PBS with 0.05% BSA
4613
IgG2a κ
Protein A or G purified
NMYC-1
Western Blot 1 to 2 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat
BHLHE37, bHLHe37N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, MODED, neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene, N-myc, NMYCneuroblastoma MYC oncogene, ODED, oncogene NMYC, pp65/67, v-myc avian myelocytomatosis viral related oncogene, neuroblastoma derived, v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian)
Recombinant full-length human n-Myc protein.
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.