missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N myc interactor Mouse anti-Human, Clone: PCRP-NMI-1C1, Novus Biologicals™
Mouse Monoclonal Antibody
508.00€
Specifications
| Antigen | N myc interactor |
|---|---|
| Clone | PCRP-NMI-1C1 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Protein Array |
| Applications | ELISA, Peptide Array |
Description
N myc interactor Monoclonal specifically detects N myc interactor in Human samples. It is validated for Flow Cytometry, Protein Array.Specifications
| N myc interactor | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Signal Transduction, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 9111 | |
| IgG2b | |
| Protein A or G purified |
| PCRP-NMI-1C1 | |
| Flow Cytometry 1 to 2 μg/million cells, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Nmi, N-myc (and STAT) interactor, N-myc and STAT interactor, N-myc interactor, N-myc-interactor | |
| Recombinant full-length human N myc interactor protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title