missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ n-Myc Antibody (PCRP-MYCN-1A9) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26972B
This item is not returnable.
View return policy
Description
n-Myc Monoclonal antibody specifically detects n-Myc in Human samples. It is validated for Flow Cytometry,ELISA
Specifications
| n-Myc | |
| Monoclonal | |
| Biotin | |
| BHLHE37, bHLHe37N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, MODED, neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene, N-myc, NMYCneuroblastoma MYC oncogene, ODED, oncogene NMYC, pp65/67, v-myc avian myelocytomatosis viral related oncogene, neuroblastoma derived, v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) | |
| Recombinant protein domain (around aa335-463) of human n-Myc protein | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 4613 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA | |
| PCRP-MYCN-1A9 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction