missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Myc Antibody (NMYC-1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-109AF594
This item is not returnable.
View return policy
Description
n-Myc Monoclonal specifically detects n-Myc in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| n-Myc | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MYCN | |
| A synthetic peptide (SPYVESEDAPPQKC) conjugated to KLH, corresponding to amino acids 327-339 of human n-Myc. [UniProtNo. P04198] | |
| Primary | |
| This antibody does not react with c-Myc or L-Myc. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| NMYC-1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunocytochemistry | |
| BHLHE37, bHLHe37N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, MODED, neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene, N-myc, NMYCneuroblastoma MYC oncogene, ODED, oncogene NMYC, pp65/67, v-myc avian myelocytomatosis viral related oncogene, neuroblastoma derived, v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) | |
| Mouse | |
| 0.1 mL | |
| 4613 | |
| Human, Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction