missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30320201
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320201 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320201 Supplier Dr. Ehrenstorfer Supplier No. DREA14195500ME1000
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material n-Hexane
CAS 110-54-3
Analyte or Component Names n-Hexane
Shipping Condition Room Temperature
Molecular Formula C6 H14
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Hexane, NSC 68472, Skellysolve B, n-Hexane
InChI Formula InChI=1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3
SMILES CCCCCC
IUPAC Name hexane
Solvent or Matrix Methanol
Molecular Weight (g/mol) 86.18
Quantity 1 mL
Formula Weight 86.1096 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.